APOH316CDE3A Apollo Scientific Ltd

2-Thienyl disulphide

  • MDL number MFCD00066333
  • Cas Number : 6911-51-9
  • Molecular Weight : 230.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 58
Iupac Name 2-(2-thienyldisulfanyl)thiophene
Inchi Code 1S/C8H6S4/c1-3-7(9-5-1)11-12-8-4-2-6-10-8/h1-6H
Inchi Key YOLFWWMPGNMXFI-UHFFFAOYSA-N

Description

Purity 95%
Flash Point 132
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 58
Product Link https://store.apolloscientific.co.uk/product/2-thienyl-disulphide

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/2-thienyl-disulphide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart