APOH316CDE3B Apollo Scientific Ltd

3-Chloro-N'-hydroxy-5-(trifluoromethyl)picolinimidamide

  • MDL number MFCD30188596
  • Cas Number : 303997-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-chloro-n-hydroxy-5-trifluoromethylpicolinimidamide

Safety Information

signalword Warning
pcodes P280;P302+P352;P362+P364
MSDS https://store.apolloscientific.co.uk/product/3-chloro-n-hydroxy-5-trifluoromethylpicolinimidamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart