APOH316CDE3E Apollo Scientific Ltd

1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone

  • MDL number MFCD12406877
  • Cas Number : 1823182-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131294

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131294
Availability details for
Quantity
  • Availability
  • Add to Cart