APOH316CDE50 Apollo Scientific Ltd

4-Cyclopropyl-2-fluoro-1-nitrobenzene

  • MDL number MFCD22418246
  • Cas Number : 932700-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at -20
Physical Form Solid-Low Melt
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-cyclopropyl-2-fluoro-1-nitrobenzene

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/4-cyclopropyl-2-fluoro-1-nitrobenzene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart