APOH316CDE51 Apollo Scientific Ltd

4-Cyclopropyl-1-fluoro-2-nitrobenzene

  • MDL number MFCD17677330
  • Cas Number : 769159-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=129411

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=129411
Availability details for
Quantity
  • Availability
  • Add to Cart