APOH316CDE57 Apollo Scientific Ltd

6-Iodo-7H-purine

  • MDL number MFCD08705723
  • Cas Number : 2545-26-8
  • Molecular Weight : 246.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-iodo-9H-purine
Inchi Code 1S/C5H3IN4/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H,7,8,9,10)
Inchi Key NIBFSSALYRLHPY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-iodo-7h-purine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-iodo-7h-purine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart