APOH316CDE61 Apollo Scientific Ltd

2-Cyclopropyl-5-fluoroaniline

  • MDL number MFCD30322597
  • Cas Number : 1538364-89-4
  • Molecular Weight : 151.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at -20
Physical Form Solid-Low Melt
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-cyclopropyl-5-fluoroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-cyclopropyl-5-fluoroaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart