APOH316CDE62 Apollo Scientific Ltd

2-Bromo-6-chloro-4-fluorobenzaldehyde

  • MDL number MFCD28733865
  • Cas Number : 1433990-64-7
  • Molecular Weight : 237.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-6-chloro-4-fluorobenzaldehyde
Inchi Code 1S/C7H3BrClFO/c8-6-1-4(10)2-7(9)5(6)3-11/h1-3H
Inchi Key YFXZCTDYQNCIEF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-chloro-4-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-chloro-4-fluorobenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart