APOH316CDE92 Apollo Scientific Ltd

8-Bromo-5-fluoroquinolin-4(1H)-one

  • MDL number MFCD15527288
  • Cas Number : 1065092-35-4
  • Molecular Weight : 242.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-5-fluoro-4(1H)-quinolinone
Inchi Code 1S/C9H5BrFNO/c10-5-1-2-6(11)8-7(13)3-4-12-9(5)8/h1-4H,(H,12,13)
Inchi Key NGLVFYGOSNNWEE-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-bromo-5-fluoroquinolin-41h-one

Safety Information

signalword Warning
pcodes P304+P340;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/8-bromo-5-fluoroquinolin-41h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart