APOH316CDEB9 Apollo Scientific Ltd

3-Fluoro-4-chlorophenacyl bromide

  • MDL number MFCD13194237
  • Cas Number : 231297-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-(4-chloro-3-fluorophenyl)ethanone
Purity 98
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=132501

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=132501
Availability details for
Quantity
  • Availability
  • Add to Cart