APOH316CDEE3 Apollo Scientific Ltd

5-Bromo-3-fluoro-2-hydrazinopyridine

  • MDL number MFCD09999913
  • Cas Number : 1289084-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=132528

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=132528
Availability details for
Quantity
  • Availability
  • Add to Cart