APOH316CDEE4 Apollo Scientific Ltd

2-Bromo-3,6-difluorobenzal fluoride

  • MDL number MFCD28166242
  • Cas Number : 1672663-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=132531

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=132531
Availability details for
Quantity
  • Availability
  • Add to Cart