APOH316CDEE5 Apollo Scientific Ltd

3-Bromo-4,5-difluorobenzal fluoride

  • MDL number MFCD28737888
  • Molecular Weight : 243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-45-difluorobenzal-fluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-45-difluorobenzal-fluoride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart