APOH316CDEE7 Apollo Scientific Ltd

5-Bromo-2,4-difluorobenzal fluoride

  • MDL number MFCD28737893
  • Cas Number : 1804408-88-5
  • Molecular Weight : 243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-5-(difluoromethyl)-2,4-difluorobenzene
Inchi Code 1S/C7H3BrF4/c8-4-1-3(7(11)12)5(9)2-6(4)10/h1-2,7H
Inchi Key XCOMHBNRSYEIQP-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-24-difluorobenzal-fluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-24-difluorobenzal-fluoride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart