APOH316CDEEF Apollo Scientific Ltd

2-Bromo-3-iodobenzal fluoride

  • MDL number MFCD18391105
  • Cas Number : 1261552-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=132543

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=132543
Availability details for
Quantity
  • Availability
  • Add to Cart