APOH316CDEF0 Apollo Scientific Ltd

2-Bromo-5-iodobenzal fluoride 250mg pack

  • MDL number MFCD18391113
  • Cas Number : 1261496-16-5
  • Molecular Weight : 332.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-iodobenzal-fluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart