APOH316CDEF1 Apollo Scientific Ltd

3-Bromo-5-iodobenzal fluoride

  • MDL number MFCD18391111
  • Cas Number : 1261476-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=132545

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=132545
Availability details for
Quantity
  • Availability
  • Add to Cart