APOH316CF7A7 Apollo Scientific Ltd

5-Chloro-7-methoxythieno[3,2-b]pyridine

  • MDL number MFCD10000846
  • Cas Number : 90690-91-8
  • Molecular Weight : 199.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-7-methoxythieno32-bpyridine

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/5-chloro-7-methoxythieno32-bpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart