APOH316CFB7C Apollo Scientific Ltd

3-(2-Chloroethyl)-1-[3-(trifluoromethyl)phenyl]urea

  • MDL number MFCD03787347
  • Cas Number : 500546-66-7
  • Molecular Weight : 266.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-chloroethyl)-N'-[3-(trifluoromethyl)phenyl]urea
Inchi Code 1S/C10H10ClF3N2O/c11-4-5-15-9(17)16-8-3-1-2-7(6-8)10(12,13)14/h1-3,6H,4-5H2,(H2,15,16,17)
Inchi Key GNGFQJFEECSJOA-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-2-chloroethyl-1-3-trifluoromethylphenylurea

Safety Information

signalword Warning
pcodes P301+P312;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/3-2-chloroethyl-1-3-trifluoromethylphenylurea
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart