APOH316CFE99 Apollo Scientific Ltd

1,3-diphenyltriaz-1-ene

  • MDL number MFCD00003021
  • Cas Number : 136-35-6
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 146
Product Link https://store.apolloscientific.co.uk/product/13-diphenyltriaz-1-ene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/13-diphenyltriaz-1-ene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart