APOH316CFE9C Apollo Scientific Ltd

Methyl 5-cyano-2-hydroxybenzoate

  • MDL number MFCD00800949
  • Cas Number : 84437-12-7
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-cyano-2-hydroxybenzoate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/methyl-5-cyano-2-hydroxybenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart