APOH316D0690 Apollo Scientific Ltd

5-Bromo-4-fluoro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD12031517
  • Cas Number : 1172067-95-6
  • Molecular Weight : 215.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-4-fluoro-1h-pyrrolo23-bpyridine

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/5-bromo-4-fluoro-1h-pyrrolo23-bpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart