APOH3258198B Apollo Scientific Ltd

1-Bromo-2-(3-bromoprop-1-ynyl)benzene

  • Cas Number : 1228107-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=137750

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=137750
Availability details for
Quantity
  • Availability
  • Add to Cart