APOH3258198C Apollo Scientific Ltd

1-Bromo-3-(3-bromoprop-1-ynyl)benzene

  • Cas Number : 1333171-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=137751

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=137751
Availability details for
Quantity
  • Availability
  • Add to Cart