APOH32581DF9 Apollo Scientific Ltd

5-Cyclopropyl-2-methyl-1H-pyrazol-3-one

  • MDL number MFCD21667125
  • Cas Number : 199125-21-8
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyclopropyl-1-methyl-1H-pyrazol-5-ol
Inchi Code 1S/C7H10N2O/c1-9-7(10)4-6(8-9)5-2-3-5/h4-5,10H,2-3H2,1H3
Inchi Key SOVRIDFTTLSXMX-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-cyclopropyl-2-methyl-1h-pyrazol-3-one

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/5-cyclopropyl-2-methyl-1h-pyrazol-3-one
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart