APOH66BABB2E Apollo Scientific Ltd

Methyl 4-chloro-1-cyclopropyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD32200419
  • Cas Number : 2407349-24-8
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-chloro-1-cyclopropyl-1H-pyrazole-3-carboxylate

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-chloro-1-cyclopropyl-1h-pyrazole-3-carboxylate

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/methyl-4-chloro-1-cyclopropyl-1h-pyrazole-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart