APOH66BABB6F Apollo Scientific Ltd

3-Bromo-4-(dimethylsulfamoyl)benzoic acid

  • MDL number MFCD11131893
  • Cas Number : 1019384-91-8
  • Molecular Weight : 308.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-dimethylsulfamoylbenzoic-acid

Safety Information

signalword Warning
pcodes P302+P352;P305+P351+P338;P362+P364
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-dimethylsulfamoylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart