APOH66BABC35 Apollo Scientific Ltd

2-Bromo-1-(2-chloropyridin-4-yl)ethanone hydrobromide

  • MDL number MFCD13192035
  • Cas Number : 569667-89-6
  • Molecular Weight : 315.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-(2-chloro-4-pyridinyl)ethanone hydrobromide
Inchi Code 1S/C7H5BrClNO.BrH/c8-4-6(11)5-1-2-10-7(9)3-5;/h1-3H,4H2;1H
Inchi Key REXIGZGIILHEHG-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-1-2-chloropyridin-4-ylethanone-hydrobromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-1-2-chloropyridin-4-ylethanone-hydrobromide
pictograms GHS05
hstatements H314;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart