APOH66BABC37 Apollo Scientific Ltd

Ethyl 3-amino-7-bromo-5-chloro-1-benzofuran-2-carboxylate

  • MDL number MFCD21609336
  • Cas Number : 2183997-56-8
  • Molecular Weight : 318.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-amino-7-bromo-5-chloro-1-benzofuran-2-carboxylate
Inchi Code 1S/C11H9BrClNO3/c1-2-16-11(15)10-8(14)6-3-5(13)4-7(12)9(6)17-10/h3-4H,2,14H2,1H3
Inchi Key SYVNMTIJYMXSNF-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-amino-7-bromo-5-chloro-1-benzofuran-2-carboxylate

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-amino-7-bromo-5-chloro-1-benzofuran-2-carboxylate
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart