APOH66BABC93 Apollo Scientific Ltd

4-(Dimethylsulfamoyl)-3-fluorobenzoic acid

  • MDL number MFCD21817646
  • Cas Number : 1534724-80-5
  • Molecular Weight : 247.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-dimethylsulfamoyl-3-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-dimethylsulfamoyl-3-fluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart