APOH9081D6B7 Apollo Scientific Ltd

Ethyl 5-oxo-3-thioxo-2H-1,2,4-triazine-6-carboxylate

  • MDL number MFCD01567017
  • Cas Number : 51101-09-8
  • Molecular Weight : 201.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-oxo-3-thioxo-2h-124-triazine-6-carboxylate

Safety Information

MSDS https://store.apolloscientific.co.uk/product/ethyl-5-oxo-3-thioxo-2h-124-triazine-6-carboxylate
Availability details for
Quantity
  • Availability
  • Add to Cart