APOH9081D6C9 Apollo Scientific Ltd

5-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD18072706
  • Cas Number : 1245645-07-1
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-iodo-124triazolo15-apyridin-2-ylamine

Safety Information

MSDS https://store.apolloscientific.co.uk/product/5-iodo-124triazolo15-apyridin-2-ylamine
Availability details for
Quantity
  • Availability
  • Add to Cart