ATE428010109 AstaTech, Inc.

6-FLUORO-3-(PIPERIDIN-4-YL)BENZO[D]OXAZOL-2(3H)-ONE HCL

  • MDL number MFCD16620950
  • Cas Number : 1134819-90-1
  • Molecular Weight : 272.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8 C
Physical Form Yellow to Brown Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=40036

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=AB7297
pictograms Exclamation Mark
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart