ATE429353853 AstaTech, Inc.

TERT-BUTYL 4-(3-HYDROXYBENZYL)PIPERAZINE-1-CARBOXYLATE

  • MDL number MFCD14635733
  • Cas Number : 500013-36-5
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8 C
Physical Form White to Yellow Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=47275

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=AB8702
pictograms Exclamation Mark
hstatements H302;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart