ATE429355705 AstaTech, Inc.

L-HISTIDINE HCL

  • MDL number MFCD00064556
  • Cas Number : 1007-42-7
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8C
Physical Form White to Yellow Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=46392

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=FD20036
pictograms Exclamation Mark
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart