ATE443546409 AstaTech, Inc.

(4-(3-METHYLUREIDO)PHENYL)BORONIC ACID PINACOL ESTER

  • MDL number MFCD06795684
  • Cas Number : 874290-99-0
  • Molecular Weight : 276.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-187 C
Purity 95
Storage temp. 2-8 C
Physical Form Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=54640

Safety Information

signalword Warning
pcodes P261;P305+P351+P338
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=71604
pictograms Exclamation Mark
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart