ATE448430542 AstaTech, Inc.

BERBERINE CHLORIDE HYDRATE

  • MDL number MFCD00149997
  • Cas Number : 68030-18-2
  • Molecular Weight : 389.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point ~ 200 C(Dec)
Purity 95
Storage temp. Room temperature
Physical Form Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=56370

Safety Information

signalword Warning
pcodes P261;P280;P301+P312;P302+P352;P305+P351+P338
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=43005
pictograms Exclamation Mark
hstatements H302;H315;H320;H335
Availability details for
Quantity
  • Availability
  • Add to Cart