ATE514314704 AstaTech, Inc.

(4-AMINO-2-NITRO-PHENYL)-CARBAMIC ACID ETHYL ESTER

  • MDL number MFCD18711762
  • Cas Number : 73895-87-1
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8 C
Physical Form Light-red to Brown Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=79232

Safety Information

signalword Warning
pcodes P261;P305+P351+P338
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=W10060
pictograms Exclamation Mark
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart