ATE514315540 AstaTech, Inc.

2-BROMO-6-(TRIFLUOROMETHYL)IMIDAZO[1,2-A]PYRIDINE

  • MDL number MFCD11840963
  • Cas Number : 1135282-92-6
  • Molecular Weight : 265.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8 C
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=80090

Safety Information

signalword Warning
pcodes P264;P270;P301+P312;P330
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=W10894
pictograms Exclamation Mark
hstatements H302;H315;H320;H335
Availability details for
Quantity
  • Availability
  • Add to Cart