ATE514316776 AstaTech, Inc.

2-BROMO-4-CHLOROTHIAZOLE-5-CARBOXYLIC ACID

  • MDL number MFCD28041028
  • Cas Number : 139670-04-5
  • Molecular Weight : 242.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room temperature
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=81336

Safety Information

signalword Warning
pcodes P261;P280;P301+P312;P302+P352;P305+P351+P338
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=Y10484
pictograms Exclamation Mark
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart