ATE517250723 AstaTech, Inc.

1-(4-IODOPHENYL)PIPERIDINE

  • MDL number MFCD22418495
  • Cas Number : 55376-34-6
  • Molecular Weight : 287.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room temperature
Physical Form Yellow to Brown Solid
Shipping Temperature Normal
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=91167

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=A10114
pictograms Exclamation Mark
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart