ATE517252620 AstaTech, Inc.

2-(3,4-DIMETHOXYPHENYL)ACETALDEHYDE

  • MDL number MFCD02261749
  • Cas Number : 5703-21-9
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20 C
Physical Form Pale-yellow to Yellow-brown Liquid
Shipping Temperature Cold box
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=89452

Safety Information

signalword Warning
pcodes P261;P280;P305+P351+P338;P304+P340;P405
MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=O10461
pictograms Exclamation Mark
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart