ATE638638651 AstaTech, Inc.

NH2-PEG2-COOH

  • MDL number MFCD20926377
  • Cas Number : 791028-27-8
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Pale-yellow to Yellow-brown Sticky Oil to Semi-Solid
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=99744

Safety Information

MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=H11015
Availability details for
Quantity
  • Availability
  • Add to Cart