ATEH03AB1EAA AstaTech, Inc.

CILASTATIN

  • MDL number MFCD27939695
  • Cas Number : 82009-34-5
  • Molecular Weight : 358.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=112632

Safety Information

MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=A10603
Availability details for
Quantity
  • Availability
  • Add to Cart