ATEH03AB2C8B AstaTech, Inc.

N-EICOSYL ACETATE

  • MDL number MFCD00056263
  • Cas Number : 822-24-2
  • Molecular Weight : 340.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form White to Yellow Solid
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=113748

Safety Information

MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=A11803
Availability details for
Quantity
  • Availability
  • Add to Cart