ATEH03AB550F AstaTech, Inc.

4-((5-CHLORO-3-FLUOROPYRIDIN-2-YL)OXY)BENZONITRILE

  • MDL number MFCD22725273
  • Cas Number : 1538764-41-8
  • Molecular Weight : 248.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form White to Yellow Solid
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=116257

Safety Information

MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=F52824
Availability details for
Quantity
  • Availability
  • Add to Cart