ATEH11AB27BF AstaTech, Inc.

4-(DIFLUOROMETHYL)BENZONITRILE

  • MDL number MFCD23843788
  • Cas Number : 55805-10-2
  • Molecular Weight : 153.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Colorless to Yellow Liquid
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=128684

Safety Information

MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=W18053
Availability details for
Quantity
  • Availability
  • Add to Cart