ATEH3031E31F AstaTech, Inc.

VERUCERFONT

  • MDL number MFCD19443727
  • Cas Number : 885220-61-1
  • Molecular Weight : 406.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form White to Yellow Solid
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=132367

Safety Information

MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=43741
Availability details for
Quantity
  • Availability
  • Add to Cart