ATEH93EDCD2B AstaTech, Inc.

TERT-BUTYL (2S,5S)-5-(HYDROXYMETHYL)-2-METHYL-PIPERIDINE-1-CARBOXYLATE

  • Cas Number : 2287346-60-3
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Colorless to Yellow Liquid
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=177258

Safety Information

MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=P20137
Availability details for
Quantity
  • Availability
  • Add to Cart