ATEH94724F70 AstaTech, Inc.

PRALSETINIB

  • MDL number MFCD31692384
  • Cas Number : 2097132-94-8
  • Molecular Weight : 533.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Pale-yellow to Yellow-brown Solid
Country of Origin CN
Product Link http://astatechinc.com/CPSResult.php?CRNO=164894

Safety Information

MSDS http://astatechinc.com/Asta_MSDS.php?Catalog=49918
Availability details for
Quantity
  • Availability
  • Add to Cart